Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth Lennox Tablet mic
Catalog No. 502904425
Encompass
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids and carbohydrates in a low-salt formulation.
Catalog No. 50-290-4425 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L7275500TAB
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids and carbohydrates in a low-salt formulation.