Learn More
Teknova LB Lennox Broth, 1kg.
SDP
Catalog No. 50842694
Encompass
1.00% Tryptone, 0.50% Yeast Extract, 0.50% Sodium Chloride, 0.48% Potassium Phosphate Dibasis
Catalog No. 50-842-694 Supplier Teknova Supplier No. L9195
1.00% Tryptone, 0.50% Yeast Extract, 0.50% Sodium Chloride, 0.48% Potassium Phosphate Dibasis