Learn More
Quality Biological Inc LB Broth(Miller) Pack 4X100ML
SDP
Supplier:  Quality Biological Inc 340004721
Encompass_Preferred
This standard bacteriological medium for the propagation of E.coli strains in suspension contains 10g of NaCl per liter.
Catalog No. 50-983-236