Learn More
MOLTOX Molecular Toxicology Inc LB (Luria-Burtani) Broth Miller, 1000 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948774
Encompass_Preferred
LB Broth Miller, also referred to as Luria-Bertani Broth, is a widely used medium in molecular biology applications for maintaining and propagating E. coli. LB Broth, Miller contains twice the amount of sodium chloride as that of the Lennox formulation which allows for the researcher to be selective of the salt content when working with a specific strain. Used as an industry standard for maintaining and cultivating recombinant strains of E. coli and for the preparation of plasmid DNA, MOLTOX offers a large selection of LB Agars and Broths with plenty of added antibiotics to choose from for a more selective media. MOLTOX does not provide microorganisms for testing. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.
Catalog No. 50-948-774 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26598
LB Broth Miller, also referred to as Luria-Bertani Broth, is a widely used medium in molecular biology applications for maintaining and propagating E. coli. LB Broth, Miller contains twice the amount of sodium chloride as that of the Lennox formulation which allows for the researcher to be selective of the salt content when working with a specific strain. Used as an industry standard for maintaining and cultivating recombinant strains of E. coli and for the preparation of plasmid DNA, MOLTOX offers a large selection of LB Agars and Broths with plenty of added antibiotics to choose from for a more selective media. MOLTOX does not provide microorganisms for testing. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.