Learn More
Quality Biological Inc LB Broth (Luria Bertani, Miller), 100 mL
SDP
Supplier:  Quality Biological Inc 340004721EA
Encompass_Preferred
Nutritionally rich media designed for growth of pure cultures of recombinant strains. ¿ 10 g/L of salt formulation ¿ The higher salt concentration (NaCl) in Bertani facilitates better transport and osmotic balance. ¿ Suitable for propagation of E. Coli and other bacteriological stocks. ¿ Recommendation: This is the best option for LB Broth is a higher concentration of sodium chloride is desired; as LB Broth, Luria-Bertani Broth has 10.0 g/L NaCl and LB Broth, Lennox contains 5.00 g/L NaCl. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Autoclave Sterilized ¿ Performance Tested ¿ pH: 7.2-7.4 ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 41106200 ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-751-7356