Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc LB Broth (Miller), Case 10 X 500 mL
SDP

Catalog No. 507517295
Encompass_Preferred

Nutritionally rich media designed for growth of pure cultures of recombinant strains. 10 g/L of salt formulation The higher salt concentration (NaCl) in Bertani facilitates better transport and osmotic balance. Suitable for propagation of E. Coli and other bacteriological stocks.

Recommendation: This is the best option for LB Broth is a higher concentration of sodium chloride is desired; as LB Broth, Luria-Bertani Broth has 10.0 g/L NaCl and LB Broth, Lennox contains 5.00 g/L NaCl.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Autoclave Sterilized
Performance Tested
pH: 7.2-7.4
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7295 Supplier Quality Biological Inc Supplier No. 340004101CS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nutritionally rich media designed for growth of pure cultures of recombinant strains. 10 g/L of salt formulation The higher salt concentration (NaCl) in Bertani facilitates better transport and osmotic balance. Suitable for propagation of E. Coli and other bacteriological stocks.

Recommendation: This is the best option for LB Broth is a higher concentration of sodium chloride is desired; as LB Broth, Luria-Bertani Broth has 10.0 g/L NaCl and LB Broth, Lennox contains 5.00 g/L NaCl.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Autoclave Sterilized
Performance Tested
pH: 7.2-7.4
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.