Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth (Miller), 500mL
Catalog No. NC1654544
Encompass
Miller’s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates.
Catalog No. NC1654544 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L2542500ML
Miller’s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates.