Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ LB Broth, Miller

Catalog No. MP091005361 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091005361 5 kg
MP091005360 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP091005361 Supplier MP Biomedicals™ Supplier No. 091005361
Add to Cart
Add to Cart

For the cultivation, propagation, and maintenance of bacteria for molecular biology applications

  • For the cultivation, propagation and maintenance of E. coli , B. subtilis, S. choleraesuis, C. glutamicum and other bacteria
TRUSTED_SUSTAINABILITY

Specifications

Appearance Fine, off-white powder
Form Powder
Media Type LB Miller
Product Type Dehydrated Culture Media
Sterility Non-sterile
Formulation Contains 10 g/L sodium
For Use With (Application) Cultivation, Progagation, and Maintenance of Bacteria for Molecular Biology Applications
Packaging Type Plastic Container
Quantity 5 kg

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.