Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth Miller Highly-ref1KG
Catalog No. 502900091
Encompass
Millers LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates.
Catalog No. 50-290-0091 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L35221KG
Millers LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates.