Learn More
Genesee Scientific Corporation LB BROTH (MILLER) MIX, 500G
SDP
Catalog No. NC1860762
Encompass
High Salt, Powder Premix.500g/Unit.1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride
Catalog No. NC1860762 Supplier Genesee Scientific Corporation Supplier No. 11118
High Salt, Powder Premix.500g/Unit.1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride