Learn More
Growcells LB BROTH MILLERS 2L EA
Catalog No. 501034796
Encompass_Preferred
Ready to use sterile bacterial culture media, sterile filtered and steam sterilized in TriForest polycarbonate bottles. Recipe/L: Tryptone 10g, Yeast Extract 5g, NaCl 10g pH 7.2
Catalog No. 50-103-4796 Supplier Growcells Supplier No. MBLE7031
Ready to use sterile bacterial culture media, sterile filtered and steam sterilized in TriForest polycarbonate bottles. Recipe/L: Tryptone 10g, Yeast Extract 5g, NaCl 10g pH 7.2