Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth, Vegitone, 500 G
Catalog No. NC0726852
Encompass
For microbiology, Synonym: Luria Bertani Broth, Vegitone, Vegitone LB Broth. LB Broth Vegitone is free of animal derived material. It is used for the cultivation and maintenance of recombinant strains of Escherichia coli and may be used for routine cultivation of not particularly fastidious microorganisms.
Catalog No. NC0726852 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 28713500GF
For microbiology, Synonym: Luria Bertani Broth, Vegitone, Vegitone LB Broth. LB Broth Vegitone is free of animal derived material. It is used for the cultivation and maintenance of recombinant strains of Escherichia coli and may be used for routine cultivation of not particularly fastidious microorganisms.