Learn More
Teknova LB Broth with Animal-Free Soytone. 500mL, Sterile.
SDP
Catalog No. 501908064
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.
This item is not returnable. View return policy
Catalog No. 50-190-8064 Supplier Teknova Supplier No. L8950
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.