Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth with agar (Lennox), 250 g
Catalog No. NC1429201
Encompass
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids and carbohydrates in a low-salt formulation.
Catalog No. NC1429201 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L2897250G
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids and carbohydrates in a low-salt formulation.