Learn More
Teknova LB Broth with 6% Sucrose without Sodium Chloride. 500mL, Sterile
SDP
Supplier:  Teknova L8610
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.
Catalog No. 50-238-8112