Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth with agar Lennox
Catalog No. 502904881
Encompass
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.
Catalog No. 50-290-4881 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L7025500TAB
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.