Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth with agar Lennox1KG
Catalog No. 502904901
Encompass
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids and carbohydrates in a low-salt formulation.
Catalog No. 50-290-4901 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L28971KG
Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids and carbohydrates in a low-salt formulation.