Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth with agar Luria l1KG
Catalog No. 502900090
Encompass
Lurias low salt LB is a microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates. The addition of agar provides a solid medium for microbial growth.
Catalog No. 50-290-0090 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L32721KG
Lurias low salt LB is a microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates. The addition of agar provides a solid medium for microbial growth.