Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Broth with agar Miller1KG
Catalog No. 502904269
Encompass
Millers LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates. The addition of agar provides a solid medium for microbial growth.
Catalog No. 50-290-4269 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L31471KG
Millers LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides amino acids water-soluble vitamins and carbohydrates. The addition of agar provides a solid medium for microbial growth.