Learn More
Teknova LB Broth with Animal-Free Soytone and Kanamycin-50. 1000mL, Sterile.
SDP
Catalog No. 502388003
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.
Catalog No. 50-238-8003 Supplier Teknova Supplier No. L8137
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.