Learn More
Teknova LB Broth with Animal-Free Soytone. 200L Bag, Sterile.
SDP
Catalog No. 502745893
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.
Catalog No. 50-274-5893 Supplier Teknova Supplier No. 21360
Add to Cart
Edge
Add to Cart
Request a Quote
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.