Learn More
Teknova LB Broth with Chloramphenicol-25. 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387292
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.
Catalog No. 50-238-7292 Supplier Teknova Supplier No. L81252PK
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.