Learn More
Teknova LB Broth with Carbenicillin-100. 1000mL, Sterile.
SDP
Supplier:  Teknova L8185
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.
Catalog No. 50-842-628