Learn More
Teknova LB Broth with Carbenicillin-50. 1000mL, Sterile.
SDP
Catalog No. 50842612
Encompass
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.
This item is not returnable. View return policy
Catalog No. 50-842-612 Supplier Teknova Supplier No. L8110
Lysogeny broth(LB) is a nutritionally rich medium that is used for the growth of bacteria. It is also known as Luria broth or Luria-Bertani broth. It continues to be one of the most common media used for maintaining and cultivating recombinant strains of Escherichia coli.