Learn More
Kd Medical Inc LB+GENT+ KAN+TET+BLUOGAL
SDP
Catalog No. 501031255
Encompass
This item has a minimum order quantity of 4 per supplier requirements. Used for cultivation of E.Coli that contains plasmids conferring Gentamycin (7ug/ml), Kanamycin (50ug/ml), and Tetracyline (10ug/ml) resistance. Bluo Gal used for blue/white screening of colonies. IPTG 1mM. Ten 100mm plates per sleeve.
Catalog No. 50-103-1255 Supplier Kd Medical Inc Supplier No. BPL2553
This item has a minimum order quantity of 4 per supplier requirements. Used for cultivation of E.Coli that contains plasmids conferring Gentamycin (7ug/ml), Kanamycin (50ug/ml), and Tetracyline (10ug/ml) resistance. Bluo Gal used for blue/white screening of colonies. IPTG 1mM. Ten 100mm plates per sleeve.