Learn More
Kd Medical Inc LB+KANAMYCIN 30UG/ML
SDP
Catalog No. 501019040
Encompass_Preferred
Used for the maintenance and propagation of E. coli, selective for kanamycin resistance. Ten 100mm plates per sleeve. Concentration of Kanamycin is 30mg/ml. Ten 100mm plates per sleeve.
Catalog No. 50-101-9040 Supplier Kd Medical Inc Supplier No. BPL2580
Used for the maintenance and propagation of E. coli, selective for kanamycin resistance. Ten 100mm plates per sleeve. Concentration of Kanamycin is 30mg/ml. Ten 100mm plates per sleeve.