Learn More
Kd Medical Inc LB+KANAMYCIN 50UG/ML 150
SDP
Catalog No. 501031318
Encompass_Preferred
Used for cultivation of E.Coli that contains plasmids conferring Kanamycin (50ug/ml) resistance. Ten 150mm plates per sleeve.
Catalog No. 50-103-1318 Supplier Kd Medical Inc Supplier No. BPP6640
Used for cultivation of E.Coli that contains plasmids conferring Kanamycin (50ug/ml) resistance. Ten 150mm plates per sleeve.