Learn More
Kd Medical Inc LB (LENNOX) 500ML BOTTLE
SDP
Catalog No. 501018953
Encompass_Preferred
LB Lennox broth is used for E.coli growth and propagation; plasmid DNA and protein production. One 500ml bottle.
Catalog No. 50-101-8953 Supplier Kd Medical Inc Supplier No. BLE3060
LB Lennox broth is used for E.coli growth and propagation; plasmid DNA and protein production. One 500ml bottle.