Learn More
Teknova LB Lennox Agar. 1kg.
SDP
Catalog No. 502388034
Encompass
1.0% Tryptone, 0.5% Yeast Extract,0.5% Sodium Chloride, 1.5% Agar
Catalog No. 50-238-8034 Supplier Teknova Supplier No. L9330
1.0% Tryptone, 0.5% Yeast Extract,0.5% Sodium Chloride, 1.5% Agar