Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ LB Lennox Agar (Capsules)

Catalog No. MP113002321 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
454 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113002321 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP113002321 Supplier MP Biomedicals™ Supplier No. 113002321
Add to Cart
Add to Cart

Offers convenience and ease of use. MP Biomedicals LB Lennox Agar (Capsules) has the same formulation as LB Lennox Agar powder with the advantage of a capsule form. This rich medium is widely used for the growth of E. coli and certified for plasmid and ot

LB Lennox Agar is a nutritionally rich medium originally developed for growth and maintenance of E. coli strains. The ultimate in ease and convenience, LB Lennox Agar in capsule form eliminates the waste, inaccuracies and mess associated with weighing bulk powder. The growth characteristics are the same as LB Lennox Agar powder.

LB Lennox Agar Medium supports a luxurious growth of E. coli cells, as tryptone and yeast extract supply essential growth factors that E. coli cells would otherwise have to synthesize. The medium also contains essential electrolytes for transport and osmotic balance, due to the NaCl component. Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in LB Lennox Agar formulations.

  • Chosen reagents are combined in the proper proportions according to the formulation and thoroughly blended for a uniform distribution
  • A sample of the blended formulation is used to prepare the agar medium
  • The combinations are mixed to dissolve components and then autoclaved at 121°C for 15 minutes
  • It is recommended to use 35 capsules per liter of purified water
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage Store at room temperature.
Description LB-agar, Lennox
Format Capsule
Formulation Typically contains 10 g Tryptone; 5 g Yeast Extract; 5 g NaCl; 15 g Agar-B. Small quantities of biological buffers have been added for proper pH adjustment.
For Use With (Application) Bacterial culture
Quantity 454 g
Regulatory Status RUO
Specimen Type E. coli
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.