Learn More
Teknova LB Lennox Agar Plates. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388052
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride,1.5% Agar
Catalog No. 50-238-8052 Supplier Teknova Supplier No. L7590
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride,1.5% Agar