Learn More
Teknova LB Lennox Agar Plates. 100mm, 35mL Fill, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388115
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 2.0% Agar, 0.5% Sodium Chloride,
Catalog No. 50-238-8115 Supplier Teknova Supplier No. L3500
1.0% Tryptone, 0.5% Yeast Extract, 2.0% Agar, 0.5% Sodium Chloride,