Learn More
Teknova LB Lennox Agar Plates. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387907
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar
Catalog No. 50-238-7907 Supplier Teknova Supplier No. L58704PK
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar