Learn More
Teknova LB Lennox Agar Plates, 20mL Fill per Plate. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388019
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar
Catalog No. 50-238-8019 Supplier Teknova Supplier No. L5199
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar