Learn More
Teknova LB Lennox Agar Plates with Ampicillin-100. 100mm, 35mL Fill, 20 Plates/Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387623
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 100g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7623 Supplier Teknova Supplier No. L35048PK
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 100g/mL Ampicillin Sodium Salt