Learn More
Teknova LB Lennox Agar Plates with Carbenicillin-100. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502388140
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-8140 Supplier Teknova Supplier No. L51924PK
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt