Learn More
Teknova LB Lennox Agar Plates with Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile 4 pack.
SDP
Catalog No. 502387711
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate
Catalog No. 50-238-7711 Supplier Teknova Supplier No. L31204PK
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate