Learn More
Teknova LB Lennox Agar Plates with Zeocin-25. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387444
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,25g/mL Zeocin (an InvivoGen's product)
Catalog No. 50-238-7444 Supplier Teknova Supplier No. L10364PK
1.0% Tryptone, 0.5% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,25g/mL Zeocin (an InvivoGen's product)