Learn More
Teknova LB Lennox Agar Plates with 2% Agar, 0.05% Glucose and Ampicillin-100. 100mm, 20 Plates/Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387677
Encompass
1.00% Tryptone, 0.50% Yeast Extract, 0.50% Sodium Chloride, 2.00% Agar, 0.05% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7677 Supplier Teknova Supplier No. 2L11254PK
1.00% Tryptone, 0.50% Yeast Extract, 0.50% Sodium Chloride, 2.00% Agar, 0.05% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Ampicillin Sodium Salt