Learn More
Kd Medical Inc LB (LENNOX) + AMP 100UG/ML
SDP
Supplier:  Kd Medical Inc BPL2330
Encompass_Preferred
LB Lennox (Ampicillin) Used for the maintenance and propagation of Escherichia coli (E.coli), selective for Ampicillin (100ug/ml) resistance. Lower NaCl formulation. Ten 100mm plates per sleeve.
Catalog No. 50-101-9001