Learn More
Kd Medical Inc LB (LENNOX)+AMP+CHLOR
SDP
Catalog No. 501019003
Encompass_Preferred
Used for the maintenance and propagation of Escherichia coli (E.coli), selective for Ampicillin (100ug/ml) and Chloramphenicol (50ug/ml) resistance. Lower NaCl formulation. Ten 100mm plates per sleeve.
Catalog No. 50-101-9003 Supplier Kd Medical Inc Supplier No. BPL2333
Used for the maintenance and propagation of Escherichia coli (E.coli), selective for Ampicillin (100ug/ml) and Chloramphenicol (50ug/ml) resistance. Lower NaCl formulation. Ten 100mm plates per sleeve.