Learn More
Teknova LB Lennox Broth. 500mL. Sterile. 8 pack.
SDP
Catalog No. 502387903
Encompass
1.0% Tryptone, 0.5% Yeast Extract,0.5% Sodium Chloride
Catalog No. 50-238-7903 Supplier Teknova Supplier No. L51508PK
1.0% Tryptone, 0.5% Yeast Extract,0.5% Sodium Chloride