Learn More
Teknova LB Lennox Broth Media, 1kg
SDP
Supplier:  Teknova L9310
Encompass
1.00% Tryptone, 0.50% Yeast Extract, 0.50% Sodium Chloride, 0.35% Potassium Phosphate Dibasic
Catalog No. 50-238-8160