Learn More
Kd Medical Inc LB(LENNOX)+KANAMYCIN 50UG/ML
SDP
Catalog No. 501031263
Encompass_Preferred
Used for cultivation of E.Coli that contains plasmids conferring Kanamycin (50ug/ml)resistance. Lower NaCl concentration. Ten 100mm plates per sleeve.
Catalog No. 50-103-1263 Supplier Kd Medical Inc Supplier No. BPL2650
Used for cultivation of E.Coli that contains plasmids conferring Kanamycin (50ug/ml)resistance. Lower NaCl concentration. Ten 100mm plates per sleeve.