Learn More
Teknova LB Low Salt Top Agar. 100mL, Sterile.
SDP
Catalog No. NC1805705
Encompass
1.00% Tryptone, 0.50% Yeast Extract, 0.50% Sodium Chloride, 0.75% Agar
This item is not returnable. View return policy
Catalog No. NC1805705 Supplier Teknova Supplier No. L7570
1.00% Tryptone, 0.50% Yeast Extract, 0.50% Sodium Chloride, 0.75% Agar