Learn More
Teknova LB Luria Agar Plates with Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Supplier:  Teknova L76054PK
Encompass
1.00% Tryptone, 0.50% Yeast Extract, 0.05% Sodium Chloride, 1.50% Agar,50g/mL Kanamycin Sulfate
Catalog No. 50-238-7625