Learn More
Teknova LB Luria Agar Plates with 1.6% Skim Milk and Spectinomycin-100. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387451
Encompass
1.00% Tryptone, 0.50% Yeast Extract, 0.05% Sodium Chloride, 1.60% Skim Milk, 1.50% Agar, 100g/mL Spectinomycin Dihydrochloride Pentahydrate
Catalog No. 50-238-7451 Supplier Teknova Supplier No. L76724PK
1.00% Tryptone, 0.50% Yeast Extract, 0.05% Sodium Chloride, 1.60% Skim Milk, 1.50% Agar, 100g/mL Spectinomycin Dihydrochloride Pentahydrate