Learn More
Crescent Chemical Co Inc LB MEDIUM POWDER 20G FOR 1L
SDP
Catalog No. 50249149
Encompass_Preferred
LB Medium, Powder 20 g/L For cultivation of E. coli in molecular biology.10 g/l Tryptone5 g/l Yeast extract5 g/l NaClFor making 1 L liquid medium suspend 20 g in 900 ml distilled water, adjust the pH to 7.0 with approximately 0.2 ml of 5N NaOH, fill up to a final v
Catalog No. 50-249-149 Supplier Crescent Chemical Co Inc Supplier No. SE48501.01
LB Medium, Powder 20 g/L For cultivation of E. coli in molecular biology.10 g/l Tryptone5 g/l Yeast extract5 g/l NaClFor making 1 L liquid medium suspend 20 g in 900 ml distilled water, adjust the pH to 7.0 with approximately 0.2 ml of 5N NaOH, fill up to a final v