Learn More
CEM Corporation Amino Acid Manifold O-ring
SDP
Supplier:  CEM Corporation 167735
Encompass
The Amino Acid Manifold O-ring is an EPDM o-ring used to seal the amino acid bottles to the manifold.
Catalog No. 50-187-7693