Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc LB Agar Plates with 50 µg/mL Kanamycin, Sleeve 10 X 25 mL
SDP

Catalog No. 507517584
Encompass_Preferred

LB Agar Plates are used for the growth and maintenance of E. coli strains used in molecular microbiology procedures.

•with 50 µg/mL Kanamycin

Applications: Nutritionally rich media designed for growth of pure cultures of recombinant strains. LB Agar plates are suitable for propagation of E. coli and other bacteriological stocks.

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Autoclave Sterilized

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7584 Supplier Quality Biological Inc Supplier No. 340147231
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LB Agar Plates are used for the growth and maintenance of E. coli strains used in molecular microbiology procedures.

•with 50 µg/mL Kanamycin

Applications: Nutritionally rich media designed for growth of pure cultures of recombinant strains. LB Agar plates are suitable for propagation of E. coli and other bacteriological stocks.

Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Autoclave Sterilized

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.